Tisk…
Work positioning lanyards, Protecta®, 3M™
Dodavatel: CAPITAL SAFETY
1200314 AL410C AL410C2 AL415C AL420C2 AL422 AL432/1
115-2672EA 0 CZK
115-2672 115-2669 CPSDAL410C2 CPSDAL415C 115-2670 115-2671 CPSDAL432/1
Work positioning lanyards, Protecta®, 3M™
Ochrana před pádem Tlumiče pádu a lana
Protecta® work positioning rope connecting lanyards are an ideal solution for the connecting element in a restraint system. Made from polyamide braided rope with safety stitching.
  • Made from Ø10,5 mm durable and flexible braided polyamide rope
  • Double-leg and single-leg formats are available
  • Sewn terminations with heat shrink tube protection provide greater durability and long product service life
  • Use as part of a restraint system to prevent the user from moving into an area where they may be exposed to a fall
They are available in a variety of lengthsMore Product Information