Tisk…
Cleanroom wipes, UltraSeal® 3000
Dodavatel: BERKSHIRE
US3000.0909.8
115-1273EA 78440 CZK
115-1273
Cleanroom wipes, UltraSeal® 3000
Ubousky/Utěrky Utěrky pro čisté prostory Ubrousky nevlhčené do čistých prostor
Tubular sealed edge 2-ply polyester wipes are ideal for cleaning critical tools and machines, designed for use in manufacturing devices sensitive to micron particle contamination, non volatile residues and ions.
  • Sealed edges, with wide heat seals, low particles and fibres due to only 2 edges
  • Tear-resistant for tough cleaning, double thickness for high absorbency
  • Work well with isopropanol and other solvents
  • Steam autoclavable
  • Lot tracing on each pack
Suitable for use in class 1, ISO 3, M1.5 cleanrooms.
Balení: Laundered and packed in an ISO 4,More Product Information