Tisk…
Cleanroom wipes, pre-saturated, sterile, SatPax® 1200
Dodavatel: BERKSHIRE
SSP120000312
115-0362EA 1850 CZK
115-0362
Cleanroom wipes, pre-saturated, sterile, SatPax® 1200
Ubousky/Utěrky Utěrky pro čisté prostory Ubrousky vlhčené do čistých prostor
Durable, 100% knitted polyester wipes for the most critical aseptic applications. Pre-saturated with 70% USP-grade isopropanol and 30% deionised water.
  • Lot number on each pack satisfies current Good Manufacturing Practices (cGMP) requirements for traceability
  • Package-tested to assure sterility and chemical integrity
  • Sterilised by gamma radiation to a Sterility Assurance Level (SAL) of 10⁻⁶ in accordance with ANSI/ISO/AAMI guidelines
  • Laundered and packaged in an ISO Class 4 (Class 10) cleanroom
  • Certificate of Sterility included in each case