Tisk…

Dusičnan stříbrný ≥99.9% (metals basis) ACS

Dodavatel: Thermo Fisher Scientific
Nebezpečí

11414.06 11414.14 11414.22 11414.36 11414.62 11414.A1
11414.06EA 1500 CZK
11414.06 11414.14 11414.22 11414.36 11414.62 11414.A1
Dusičnan stříbrný ≥99.9% (metals basis) ACS
Dusičnan stříbrný

Purity: 99.9+% CAS No.: 7761-88-8 Molecular Formula: AgNO3 Formula Weight: 169.87 Physical Form: Crystalline Density: 4.352 Appearance: White Odour: Odourless Melting Point: 212° Solubility: Freely soluble in water, alcohol, ammonia water. Slightly soluble in ether Merck Reference: 14,8518 UN No.: 1479 MDL No.: MFCD00003414


Silver plating, photography, manufacturing of other silver compounds, mirrors, as an analytical lab reagent, in Colouring porcelain

Vzorec: AgNO₃
Bod varu: 444 °C (1013 hPa)
Bod tání: 212 °C
Hustota: 4,352
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS číslo: 7761-88-8
EINECS: 231-853-9
UN: 1479
ADR: 5.1,I
REACH: 01-2119513705-43
Merck Index: 14,8518
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR