Tisk…
Tychem® 6000 AL TF640 WG Ventilated Coveralls
Dodavatel: DuPont
TF0640TGYWG/2X TF0640TGYWG/3X TF0640TGYWG/L TF0640TGYWG/M TF0640TGYWG/S TF0640TGYWG/XL
114-9625EA 16270 CZK
114-9625 114-9626 114-9623 114-9622 114-9621 114-9624
Tychem® 6000 AL TF640 WG Ventilated Coveralls
Jednorázové oděvy
Air-fed coverall with attached non-dissipative under gloves and over boot assembly, consisting of integrated dissipative inner boots attached to dissipative outer boots.
  • Ventilated coverall with stitched and overtaped seams
  • Fully enclosed over hood with clear front and crown see-through visor
  • Front closure with double flap hook and loop, single zipper closing to center, horizontal opening
  • Three exhaust valves; two at hips, one in hood back
  • Air distribution made of Tychem 6000 fabric
  • Emergency egress
  • Attached non dissipative undergloves
  • Over boot assembly
More Product Information