Tisk…
Cleanroom Overalls, CleanMax®
Dodavatel: LAKELAND CLEANMAX APPAREL
CTL417CM-2X CTL417CM-3X CTL417CM-4X CTL417CM-5X CTL417CM-LG CTL417CM-MD CTL417CM-XL
114-9584EA 11340 CZK
114-9584 114-9585 114-9586 114-9587 114-9588 114-9589 114-9590
Cleanroom Overalls, CleanMax®
Jednorázové oděvy
Clean manufactured disposable coveralls are individually packaged and folded to promote easier gowning with less excursions.
  • Clean manufactured in silicone-free environment
  • Latex-free
  • Tunnelled elastic on wrists with thumb loops
  • Zipper closure
  • For critical environments
  • Individually bagged
CleanMax® cleanroom garments provide comfort, quality, and protection.
All these apparels provide chemical penetration resistance to oils, bleach, and 50% sodium hydroxide. CleanMax® apparel is also resistant to blood and body fluid penetration, viral penetration,More Product Information