Tisk…
Cleanroom frocks/lab coats, CleanMax®
Dodavatel: LAKELAND CLEANMAX APPAREL
CTL191CM-2X CTL191CM-3X CTL191CM-4X CTL191CM-5X CTL191CM-LG CTL191CM-MD CTL191CM-XL
114-9563EA 10540 CZK
114-9563 114-9564 114-9565 114-9566 114-9567 114-9568 114-9569
Cleanroom frocks/lab coats, CleanMax®
Jednorázové oděvy
Clean manufactured disposable frocks and lab coats are individually bagged and folded to promote easier gowning with less excursions.
  • Clean manufactured in silicone-free environment
  • Latex-free
  • Mandarin collar
  • Tunnelled elastic wrists with thumb loops
  • Zipper closure
  • No pockets
  • Individually bagged
CleanMax® cleanroom garments provide comfort, quality, and protection.
All these apparels provide chemical penetration resistance to oils, bleach, and 50% sodium hydroxide. CleanMax® apparel is also resistant to blood and body fluid penetration, viralMore Product Information