Tisk…
ESD safety shoes, lace-up, X-Light, 7131033/7131038
Dodavatel: Abeba
7131033/35 7131033/36 7131033/37 7131033/38 7131033/39 7131033/40 7131033/41 7131033/42 7131033/43 7131033/44 7131033/45 7131033/46 7131033/47 7131033/48 7131038/35 7131038/36 7131038/37 7131038/38 7131038/39 7131038/40 7131038/41 7131038/42 7131038/43 7131038/44 7131038/45 7131038/46 7131038/47 7131038/48
114-8538EA 3100 CZK
114-8538 114-8539 114-8540 114-8541 114-8542 114-8543 114-8544 114-8545 114-8546 114-8547 114-8548 114-8549 114-8550 114-8551 ABEB7131038/35 ABEB7131038/36 ABEB7131038/37 ABEB7131038/38 ABEB7131038/39 ABEB7131038/40 ABEB7131038/41 ABEB7131038/42 ABEB7131038/43 ABEB7131038/44 ABEB7131038/45 ABEB7131038/46 ABEB7131038/47 ABEB7131038/48
ESD safety shoes, lace-up, X-Light, 7131033/7131038
Obuv
ESD safety shoe made from smooth leather, reflective inserts, and a PU sole which is resistant to fuels.
  • Perspiration-absorbent, Silver Point lining
  • Replaceable active comfort insole provides optimised comfort; orthopaedic insoles possible
  • Sanitized® treatment reduces odour and bacterial growth
  • Lightweight PU sole with high slip resistance and shock absorbing system
  • Protective steel toecap (200 J)
Colour: White/black
EN 61340 (ESD), EN ISO 20345:2011; S2, SRA
Certifikace: EN 61340 (ESD); DGUV 112-191; EN ISO 20345:2011; S2, SRC.