Tisk…
Safety shoes, lace-up, Caleo, sperian
Dodavatel: HONEYWELL SAFETY
6246101/48 6246101/49 6246101/44 6246101/47 6246101/46 6246101/39 6246101/40 6246101/41 6246101/42 6246101/43 6246101/45
114-3728EA 3260 CZK
114-3728 114-3729 114-3724 114-3727 114-3726 114-3719 114-3720 114-3721 114-3722 114-3723 114-3725
Safety shoes, lace-up, Caleo, sperian
Obuv
Water resistant, oiled, full grain leather upper with reinforced protection at the front and back. Padded ankle collar and tongue, lateral watertight bellows, 3-D lining plus dual-density PU outsole (resistant to changes in temperature, abrasion, acids and alkalis in limited to low concentrations).
  • Light and flexible PU outsole ensures shock absorption and high slip resistance
  • Antibacterial lining with 3-D structure and textile panels
  • Removable, dual-density PU foam insole; antibacterial, antistatic and with comfortable heel pads
  • Light, metal-free Spacium™
More Product Information