Tisk…
Safety shoes, lace-up, The Roar, King
Dodavatel: U-Power
SN20014/38 SN20014/39 SN20014/40 SN20014/41 SN20014/42 SN20014/43 SN20014/44 SN20014/45 SN20014/46 SN20014/47 SN20014/48 SN20014/35 SN20014/36 SN20014/37
114-2867EA 3220 CZK
114-2867 114-2868 114-2869 114-2870 114-2871 114-2872 114-2873 114-2874 114-2875 114-2876 114-2877 114-2864 114-2865 114-2866
Safety shoes, lace-up, The Roar, King
Obuv
Safety shoes made from soft water repellent leather and a slip-resistant dual-density PU sole.
  • Light non-slip Action dry insole made from EVA/textile keeps feet dry; anti-fungal treated
  • Wingtex tunnel lining with honeycomb structure ensures breathability and excellent absorbance features
  • Natural fit width 11 for additional comfort
  • Air toe protective aluminium toecap with breathable membrane (200 J)
  • Light, flexible and non-metallic puncture resistant midsole 'Save & Flex' covers 100% of the sole of the foot
Colour: Black
Colour: Black