Tisk…

Dusičnan ceričito-amonný ≥99.5% (REO, rare earth oxide basis), REacton®

Dodavatel: Thermo Fisher Scientific
Nebezpečí

11332.09 11332.22 11332.36
11332.09EA 869 CZK
11332.09 11332.22 11332.36
Dusičnan ceričito-amonný ≥99.5% (REO, rare earth oxide basis), REacton®
Dusičnan ceričito-amonný

Purity: 99.5% CAS No.: 16774-21-3 Molecular Formula: H8CeN8O18 Formula Weight: 548.23 Physical Form: Crystalline Appearance: Orange Odour: Pungent Merck Reference: 14,1992 UN No.: 1477 MDL No.: MFCD00151121

Vzorec: (NH₄)₂Ce(NO₃)₆
Bod tání: 107…108 °C
Hustota: 2,2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00151121
CAS číslo: 16774-21-3
EINECS: 240-827-6
UN: 1477
ADR: 5.1,II
Merck Index: 14,1992

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR