Tisk…
Cleanroom overalls, BioClean-D™ CleanTough™, disposable
Dodavatel: Nitritex
S-BDCHT-XL S-BDCHT-L S-BDCHT-M S-BDCHT-XS BDCHT-XS S-BDCHT-XXL BDCHT-L BDCHT-XL BDCCT-S BDCCT-XL BDCCT-M BDCCT-L BDCCT-XXL S-BDCCT-L S-BDCCT-S S-BDCCT-2XL S-BDCCT-M S-BDCCT-XL BDCHT-XXL BDCHT-S BDCHT-M S-BDCHT-S BDCCT-XXXL S-BDCHT-XXXL BDCHT-XXXL BDCHT-XXXXL S-BDCHT-XXXXL
113-8019EA 8780 CZK
113-8019 113-8016 113-8017 113-8032 113-8008 113-8033 113-8004 113-8007 113-7997 113-7998 113-7996 113-7995 113-7994 113-8010 113-8012 113-8009 113-8011 113-8013 113-8000 113-8006 113-8005 113-8018 113-8196 113-8365 114-9441 114-9442 NITRS-BDCHT-XXXXL
Cleanroom overalls, BioClean-D™ CleanTough™, disposable
Jednorázové oděvy
These disposable overalls made from CleanTough™ fabric provide good protection against hazardous dusts (including nuclear particles) and limited protection against chemical splashes. Available with hood or with collar.
  • Lightweight, low linting and hard-wearing material
  • Available as sterile or non sterile version
  • Zip front with flap cover, thumb loops
  • Elasticated back, cuffs and ankles
  • Each size carries a unique barcode on the outer carton (ITF-14), this allows automatic data entry into stock control systems
Suitable for use in Class 10, ISO 4, M2.5More Product Information