Tisk…
Flame-retardant overalls, Bizweld BIZ 1
Dodavatel: Portwest
BIZ1BGRL BIZ1BKRL BIZ1BKRM BIZ1BKRS BIZ1BKRXL BIZ1BKRXXL BIZ1BKRXXXL BIZ1ORRM BIZ1ORRS BIZ1ORRXL BIZ1ORRXXL BIZ1ORRXXXL BIZ1RBRL BIZ1RBRM BIZ1RBRS BIZ1RBRXL BIZ1RBRXXL BIZ1RBRXXXL BIZ1RERL BIZ1RERM BIZ1RERS BIZ1RERXL BIZ1RERXXL BIZ1RERXXXL BIZ1NARL BIZ1NARM BIZ1NARS BIZ1NARXL BIZ1NARXXL BIZ1NARXXXL BIZ1ORRL BIZ1NARXS
113-6942EA 2330 CZK
113-6942 113-6948 113-6949 113-6950 113-6951 113-6952 113-6953 113-6967 113-6968 113-6969 113-6970 113-6971 113-6972 113-6973 113-6974 113-6975 113-6976 113-6977 113-6978 113-6979 113-6980 113-6981 113-6982 113-6983 113-6960 113-6961 113-6962 113-6963 113-6964 113-6965 113-6966 PRTWBIZ1NARXS
Flame-retardant overalls, Bizweld BIZ 1
Pracovní oděvy
This flame-retardant boilersuit is most suitable for use within the welding industry, with excellent protection for workers who are exposed to extreme heats. Made of Bizweld 330 fabric (100% cotton drill).
  • Generous fit provides comfort and allows the wearer to work unhindered
  • Side pockets with access, rule pocket, two chest pockets with flaps, inner mobile phone pocket
  • Concealed press stud front
Available in many different colours.
Certifikace: EN ISO 11611 (Class 1, A1+A2), EN ISO 11612 (A1+A2, B1, C1, F1)