Tisk…
High visibility rain trousers, Bastogne 6361
Dodavatel: SIOEN
6361/FC1/S 6361/FC1/M 6361/FC1/L 6361/FC1/1XL 6361/FC1/2XL 6361/FY1/S 6361/FY1/M 6361/FY1/L 6361/FY1/1XL 6361/FY1/2XL 6361/FY1/3XL 6361/FC1/3XL
113-5978EA 1250 CZK
113-5978 113-5977 113-5976 113-5982 113-5983 113-5981 113-5980 113-5979 113-5984 113-5985 113-5975 SIOE6361/FC1/3XL
High visibility rain trousers, Bastogne 6361
Pracovní oděvy
Rain trousers with elasticated waist and high frequency welded seams. Made of Flexothane® Essential (100% polyester knitting with PU coating).
  • Waterproof and windproof
  • Legs adjustable by press studs
  • With reflective tape
Weight: 170 g/m²
Certifikace: EN 471 (1-2), ENV 343 (3-1)