Tisk…
Chemical resistant overalls, Tychem® 2000 C, models CHA5 / CHA6
Dodavatel: DuPont
TCCHA5TYLLG002 TCCHA5TYLMD002 TCCHA5TYLSM002 TCCHA5TYLXL002 TYCCHA5TYL16/L TYCCHA5TYL16/M TCCHA5TYL2X002 CHA5TYL16/XL CHA5TYL16/XXL CHA5TYL16/XXXL TCCHA5TYL3X002
113-0495EA 26050 CZK
113-0495 113-0496 113-0497 113-0498 113-0499 113-0529 113-0485 113-0486 113-0487 113-0488 113-3891
Chemical resistant overalls, Tychem® 2000 C, models CHA5 / CHA6
Jednorázové oděvy
Made from a lightweight and durable fabric (<450 g per garment), Tychem® 2000 C garments utilise the strength of Tyvek®, and a polymeric barrier coating, to offer good permeation barrier protection against a wide range of inorganic chemicals and biological hazards (even under pressure).
  • Hood; elasticated face, wrists, waist and ankles for an optimal fit
  • Thumb loops prevent sleeves from riding up
  • Self-adhesive double-zip flap and chin flap for tight seal of suit to face masks
  • Stitched, over-taped seams with barrier tape for protection and strength
  • Bright
More Product Information
Objednat
Produktová specifikace
Environmentally Preferable
Antistatický +
Rod
Aplikace oblečení Chemical protection,Laboratory,Industrial,Food
Hmotnost 83,2 g/m²