Tisk…
Shoe covers, Tyvek® 500, models POS0 / POSA
Dodavatel: DuPont
TYPOS0SWH00040 TYVPOSASWH3602 TYVPOSASWH4202
113-0491EA 11710 CZK
113-0491 113-0492 113-0493
Shoe covers, Tyvek® 500, models POS0 / POSA
Jednorázové oděvy
Specially designed for use with Tyvek® apparel, these Tyvek® 500 shoe covers can help offer enhanced protection for body parts that are more exposed to hazardous substances.
  • Ultra-low linting fabric
  • Elasticated ankle
  • Stitched internal seams
  • Anti-static treatment (EN 1149-5) on both sides (excludes slip-retardant sole)
  • Two styles available: with or without slip-retardant sole
Tyvek® is composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort. The fabric is permeable to both air and water vapour,More Product Information