Tisk…
Kimtech™ G3 White Nitrile Gloves
Dodavatel: KIMBERLY CLARK
56880 56881 56882 56883 56886
112-6402EA 8680 CZK
112-6402 112-6401 112-6400 112-6399 112-6398
Kimtech™ G3 White Nitrile Gloves
Rukavice
Kimtech™ G3 white nitrile ambidextrous gloves deliver enhanced contamination protection and tactile performance in sensitive applications. The powder-free gloves are static dissipative and feature a high grip for delicate, non sterile processes.
  • Certificate of analysis available online
  • Nitrile construction is stronger and leaner than latex gloves
  • Optimal protection against a wide range of contaminants including micro-organisms, viruses, and chemical splash
Kimtech™ G3 white nitrile gloves are ideal for handling delicate equipment in microelectronics,More Product Information