Tisk…
Nitrile gloves, Showa 707HVO
Dodavatel: Showa
707HVO-10/XL 707HVO-11/XXL 707HVO-7/S 707HVO-8/M 707HVO-9/L
112-6105EA 1040 CZK
112-6105 112-6106 112-6108 112-6109 112-6110
Nitrile gloves, Showa 707HVO
Rukavice
The 707HVO gloves are a more environmentally-conscious choice, breaking down by 82% in just 386 days when tested in a laboratory.
  • Eco-friendly alternative to standard single-use gloves
  • Highly tactile and fit like a second skin
  • Increased visibility thanks to fluorescent orange colour
  • Rolled cuff for easier donning
  • Provide protection against grease, chemicals, and abrasion
  • Ideal for use when working in laboratories or food preparation zones
Recommended for: Chemical spray and treatment; coating preparation; food packing and analysis; pharmaceuticals andMore Product Information