Tisk…
Latex gloves, TouchNTuff® 69-318
Dodavatel: Ansell
69-318/7.5-8 69-318/8.5-9 69-318/9.5-10 69-318/5.5-6 69-318/6.5-7
112-4264EA 1080 CZK
112-4264 112-4265 112-4266 112-4262 112-4263
Latex gloves, TouchNTuff® 69-318
Rukavice
These ambidextrous gloves are resistant to splashes of acids, bases, alcohols and dilute aqueous solutions of most chemicals.
  • Flexible and comfortable to wear
  • Cooler than vinyl, with superior finger flexibility and grip
  • Available in both powdered and powder-free versions
  • Powdered gloves have a smooth surface, powder-free gloves have a textured finish
Ideal for use in laboratory analysis, electronic manufacturing and in pharmaceutical industries.
EN 374, Cat. III; AQL 1,5 Food contact approved
Certifikace: EN ISO 374-1:2016 Type B (KPT), EN ISO 37More Product Information
Objednat
Produktová specifikace
Model 69-318
Environmentally Preferable
Obouruké/jednoruké Obouruké
Jednorázové/znovupoužitelné Jednorázové
Pudrované/nepudrované Nepudrované
Provedení Textured
Typ manžety Rolovaná
Norma EN EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUS
Schváleno pro styk s potravinami +