Tisk…
Isolator gloves, AlphaTec® latex
Dodavatel: Ansell
55-107/10 55-104/10 55-109/10 55-100/10 55-100/8 55-100/9 55-101/10 55-101/8 55-101/9 55-105/10 55-110/10
112-3447EA 5560 CZK
112-3447 112-3441 112-3451 112-3431 112-3432 112-3433 112-3434 112-3435 112-3436 112-3443 112-3452
Isolator gloves, AlphaTec® latex
Rukavice
These ambidextrous gauntlets are made ​​of natural rubber latex without an inner lining. They provide protection against chemical and mechanical hazards as well as against biohazards. For use with most available cleanroom glove boxes.
  • Highly resistant to most acids, alkalis, alcohols, aldehydes and ketones
  • Good tear resistance
  • Beaded cuffs for easier donning
  • Liquid-tight
Ideal for chemical and medical research, electronics, pharmaceuticals, sandblasting, and steam cleaning.
Colour: Natural
EN 388, EN 374 (AKL); Cat. III
Certifikace: EN ISO 374More Product Information