Tisk…

Aluminium bromide ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Aluminum bromide

11113.09 11113.22 11113.59
11113.09EA 1160 CZK
11113.09 11113.22 11113.59
Aluminium bromide ≥98%
Aluminium bromide

Purity: 98% CAS No.: 7727-15-3 Molecular Formula: AlBr3 Formula Weight: 266.71 Physical Form: Crystalline Density: 3.21 Appearance: White to orange Odour: Pungent Melting Point: 97.5° Boiling Point: 265° Solubility: Reacts with water violently, soluble in alcohol, carbon disulfide, ether Merck Reference: 14,332 UN No.: 1725 MDL No.: MFCD00003421


Alkylation, bromination and isomerization catalyst in organic synthesis

Vzorec: AlBr₃
MW: 266.69
Bod varu: 265 °C
Bod tání: 97.5 °C
Hustota: 3.21
Storage Temperature: Ambient
MDL Number: MFCD00003421
CAS číslo: 7727-15-3
EINECS: 231-779-7
UN: 1725
ADR: 8 II
Merck Index: 13,00330

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR