Tisk…
Goggles, uvex megasonic
Dodavatel: UVEX
9320.265 9320.281
111-2544EA 969 CZK
111-2544 UVEX9320.281
Goggles, uvex megasonic
Brýle bezpečnostní
Megasonic goggle's revolutionary lens design does not restrict the wearer's field of vision, meaning users have the clearest possible view at all times, even under difficult conditions. It has a sporty, ergonomic design and is extremely comfortable to wear, making it a reliable piece of equipment for everyday work.
  • Unique frameless lens design
  • OTG compatible with frame recess for side arms
  • Soft, adaptive face seal for incredible comfort and fit
  • 30 mm wide textile headband for a superior, no pressure fit
  • Easily interchangeable headband and lens
  • Highly
More Product Information