Tisk…
Safety spectacles, SecureFit™ 400X series
Dodavatel: 3M
SF401XSGAF-BLU SF401XSGAF-GRN
111-2504EA 555 CZK
111-2504 111-2505
Safety spectacles, SecureFit™ 400X series
Brýle
SecureFit™ 400X safety glasses feature 3M™ Pressure Diffusion Temple Technology, wide flexible temple arms, and a combined soft nose bridge and brow guard. These lightweight safety glasses feature a clear, anti-scratch and superior anti-fog, rimless polycarbonate lenses.
  • Safety glasses with 3M™ Pressure Diffusion Temple Technology for comfort
  • Wrap-around design improves the security of fit and prevents slipping
  • Padded temple touch points for increased comfort over the ears
  • Soft nose bridge for a more comfortable fit
  • Contoured brow guard provides additional
More Product Information