Tisk…
RD90 Respirator filters for X-plore® masks
Dodavatel: Dräger
6738811 6738771 6738770 6738810 6738812
DRAE6738811EA 3090 CZK
DRAE6738811 DRAE6738771 DRAE6738770 DRAE6738810 111-2427
RD90 Respirator filters for X-plore® masks
Polomasky a filtrační respirátory Polomasky filtrační opakovatelně použitelné Filtry pro masky
This series of filters with Dräger specific thread connection Rd90 (91 mm diameter) was designed for use in half masks. Suitable for Dräger half masks X-plore® 4390 and X-plore® 4790, via adapter 40/90 also for all masks with connection as per EN 148-1.
  • Compact filters with aluminium housing to ensure reliable recognition of possible filter damage
  • Individually packaged for optimum protection of the unused filter
  • Shelf life (from date of production): gas and combination filters 6 years, particle filters 12 years
Certifikace: EN 141:2000 re. EN 14387, 143:2000); SAI Global-approved