Tisk…
Safety spectacles, 200 series, anti-scratch, SecureFit™
Dodavatel: 3M
SF201AS
111-1610EA 307 CZK
111-1610
Safety spectacles, 200 series, anti-scratch, SecureFit™
Brýle
The SecureFit™ 200 Series are lightweight safety glasses with 3M™ pressure diffusion temple technology. These glasses feature a wraparound design that's secure and comfortable. They are available in various lens tints.
  • Economical lightweight (18 g) safety glasses
  • Innovative SecureFit™ pressure diffusion temple technology design features slotted ribs that provide comfortable pressure equalisation
  • Temples flex and self-adjust to the size of wearer's head
  • Flat and flexible temples combine easily with earmuffs
  • Easy to clean
  • Foam gasket available to help limit
More Product Information