Tisk…
Safety spectacles, SecureFit™ 200
Dodavatel: 3M
SF202AF SF201AF SF203AF
111-1593EA 0 CZK
111-1593 111-1592 111-1594
Safety spectacles, SecureFit™ 200
Brýle
Stylish, lightweight designed safety glasses features proprietary 3M™ Pressure Diffusion Temple Technology that helps diffuse pressure over the ear to enhance frame comfort while not compromising the security of fit across a diverse workforce.
  • 3M Pressure Diffusion Technology allows temples to naturally adjust to individual head sizes
  • Self-adjusting feature eliminates need to stock multiple frame sizes at worksites
  • Secure, snug fit keeps eyewear in place without slippage even when the person is in motion
Weight: 18 g
Certifikace: EN 166.