Tisk…
Filter with RD 40 connection and plastic housing
Dodavatel: HONEYWELL SAFETY
1788155 1788005 1788161 1788000 1788010 1788020 1788166 1788015 1788070 1788075 1786110 1788025
111-0566EA 950 CZK
111-0566 111-1478 111-1485 111-0562 111-1479 111-0563 DALL1788166 111-1480 111-0564 111-0565 111-1476 111-1481
Filter with RD 40 connection and plastic housing
Polomasky a filtrační respirátory Polomasky filtrační opakovatelně použitelné Filtry pro masky
Plastic filter for protecting against gases, vapours and particles; for use with Honeywell full face masks.
  • Housing made from durable plastic for a prolonged service life
  • Easy to dispose of (non metallic)
  • Lightweight and therefore particularly comfortable
  • Impregnated activated carbon from coconut shells
  • Extremely absorbent
The fitted full face mask may only be used if the concentration of contaminants corresponds to the standard for the filter being used. For use only in sufficiently ventilated areas with an oxygen volume of at least 17%.