Tisk…
Filter with RD 40 connection and aluminium housing
Dodavatel: HONEYWELL SAFETY
1783010 1783000 1785030 1786730 1786000 1785019 1785018 1785014 1785025 1785040 1783030 1783080 1785012
111-1464EA 851 CZK
111-1464 111-1463 111-0073 111-1477 111-0561 111-1473 111-1472 111-1471 111-1474 111-1475 111-1465 111-1467 111-1470
Filter with RD 40 connection and aluminium housing
Polomasky a filtrační respirátory Polomasky filtrační opakovatelně použitelné Filtry pro masky
Aluminium filter for protecting against gases, vapours and particles; for use with Honeywell full face masks.
  • Lightweight and durable aluminium housing with plastic cover
  • Unobstructed field of vision
  • RD 40 connection with DIN thread for a secure and exceptionally tight fit
  • Impregnated activated carbon from coconut shells
  • Extremely absorbent
The fitted full face mask may only be used if the concentration of contaminants corresponds to the standard for the filter being used. For use only in sufficiently ventilated areas with an oxygen volume of at leastMore Product Information