Tisk…
Full face masks, Panoramasque
Dodavatel: HONEYWELL SAFETY
1710987
111-1455EA 11060 CZK
111-1455
Full face masks, Panoramasque
Polomasky a filtrační respirátory Polomasky filtrační opakovatelně použitelné Masky celoobličejové
Panoramasque is a full face mask dedicated to offer respiratory protection, depending on the canister type assembled on it. For optimal protection, it is recommended to use aluminium or plastic Honeywell (Sperian) filters. They can only be used in sufficiently ventilated premises where the atmosphere contains a minimum oxygen volume of 17%.
  • Wide field of vision ensuring excellent lateral and downwards visibility
  • Skirt sealing flange for comfortable and tight seal
  • EPDM faceblank, making the mask lightweight and resistant
  • Good facial compatibility, one size
More Product Information