Tisk…
Respirator filters for masks, 7000 and 9000 Series, EasyLock®
Dodavatel: Moldex
9600 9030 9020 930001 9120 9230 9100 9200 9400 9500 9430 9730 9022 9032 9800
111-0699EA 3960 CZK
111-0699 111-0702 111-0703 111-1186 111-1183 111-1185 111-0701 111-1184 111-0700 111-1189 111-1187 111-1190 111-1181 111-1182 111-2305
Respirator filters for masks, 7000 and 9000 Series, EasyLock®
Polomasky a filtrační respirátory Polomasky filtrační opakovatelně použitelné Filtry pro masky
EasyLock® filters protect against gas, vapour and dust. Suitable for series 7000 and 9000 masks.
  • Bayonet connector
  • Particulate filters can either be connected with the gas cartridges or directly with the mask body
  • Adaptors and pre-filters are not necessary
  • Simpler logistics - 30% less parts
  • Particulate filters with pleated filter technology: Significantly reduces breathing resistance, improves wearer comfort and reduces fatigue
Certifikace: EN 143:2000+A1:2006 (particulate filters)
EN 14387:2004+A1:2008 (gas filters and pre-assembled filters)