Tisk…

Bromovodík 48% (w/w) vodný roztok , 99,9999% (metals basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Hydrogen bromide

10991.AA 10991.AE 10991.AP
10991.AAEA 992 CZK
10991.AA 10991.AE 10991.AP
Bromovodík 48% (w/w) vodný roztok , 99,9999% (metals basis)
Bromovodík

Purity: 99.9999% CAS No.: 10035-10-6 Molecular Formula: HBr Formula Weight: 80.92 Physical Form: Liquid Density: 1.490 Appearance: Colourless to faint yellow Beilstein Reference: 3587158 Merck Reference: 14,4778 UN No.: 1788 MDL No.: MFCD00011323

Vzorec: HBr
MW: 80.91
Bod varu: -66.7
Bod tání: -86
Hustota: 1.49
Storage Temperature: Ambient
MDL Number: MFCD00011323
CAS číslo: 10035-10-6
EINECS: 233-113-0
UN: 1788
ADR: 8 II
Merck Index: 14,4778

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR