Tisk…

Zlato ≥99.995% (metals basis), wire, Premion®, Ø 0.05 mm (0.002 in)

Dodavatel: Thermo Fisher Scientific

10969.G5 10969.G9 10969.H4
10969.G5EA 6040 CZK
10969.G5 10969.G9 10969.H4
Zlato ≥99.995% (metals basis), wire, Premion®, Ø 0.05 mm (0.002 in)
Zlato

Purity: 99.995% CAS No.: 7440-57-5 Physical Form: ≈0.04g/m Odour: Odourless MDL No.: MFCD00003436

Vzorec: Au
Bod varu: 2800 °C (1013 hPa)
Bod tání: 1063 °C
Hustota: 19,3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS číslo: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR