Tisk…

Kobalt ≥99.995% (metals basis), wire, Puratronic®, Ø 0.1 mm (0.004 in)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

10949.G1 10949.G5
10949.G1EA 2700 CZK
10949.G1 10949.G5
Kobalt ≥99.995% (metals basis), wire, Puratronic®, Ø 0.1 mm (0.004 in)
Kobalt

Purity: 99.995% CAS No.: 7440-48-4 Physical Form: ≈0.07g/m Odour: Odourless MDL No.: MFCD00010935

Vzorec: Co
Bod varu: 2870 °C (1013 hPa)
Bod tání: 1495 °C
Hustota: 8,9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010935
CAS číslo: 7440-48-4
EINECS: 231-158-0
Merck Index: 12,02488

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR