Tisk…

stříbro ≥99.999% (metals basis), powder, Premion® -22 mesh

Dodavatel: Thermo Fisher Scientific

10756.06 10756.14 10756.22
10756.06EA 2910 CZK
10756.06 10756.14 10756.22
stříbro ≥99.999% (metals basis), powder, Premion® -22 mesh
stříbro

Purity: 99.999% CAS No.: 7440-22-4 Odour: Characteristic UN No.: 3077 MDL No.: MFCD00003397

Vzorec: Ag
Bod varu: 2212 °C (1013 hPa)
Hustota: 10,5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS číslo: 7440-22-4
EINECS: 231-131-3
UN: 3077
ADR: 9,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR