Tisk…

Copper(II) nitrate hydrate ≥99.999% (metals basis), Puratronic®

Dodavatel: Thermo Fisher Scientific
Nebezpečí

10699.06 10699.14 10699.22 10699.36
10699.06EA 944 CZK
10699.06 10699.14 10699.22 10699.36
Copper(II) nitrate hydrate ≥99.999% (metals basis), Puratronic®
Copper(II) nitrate hydrate

Purity: 99.999% CAS No.: 13778-31-9 Molecular Formula: CuN2O6.xH2O Formula Weight: 187.55(anhy) Storage Temperature: 2°C to 8°C Physical Form: Crystalline Aggregates Sensitivity: Hygroscopic Appearance: Blue Odour: Odourless Merck Reference: 14,2643 UN No.: 3085 MDL No.: MFCD00149669

Vzorec: CuN₂O₆·H₂O
Storage Temperature: Refrigerator
MDL Number: MFCD00149669
CAS číslo: 13778-31-9
EINECS: 221-838-5
UN: 3085
ADR: 5.1,III
Merck Index: 14,2643

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR