Tisk…

Síran kobaltnatý heptahydrát ≥99.999% (metals basis), Puratronic®

Dodavatel: Thermo Fisher Scientific
Nebezpečí

10696.14 10696.22
10696.14EA 7350 CZK
10696.14 10696.22
Síran kobaltnatý heptahydrát ≥99.999% (metals basis), Puratronic®
Síran kobaltnatý heptahydrát

Purity: 99.999% CAS No.: 10026-24-1 Molecular Formula: CoO4S.7H2O Formula Weight: 281.10 (154.99anhy) Physical Form: Crystalline Aggregates Density: 2.03 Appearance: Dark red Odour: Odourless pH: 4 Merck Reference: 14,2448 UN No.: 3077 MDL No.: MFCD00149658

Vzorec: CoSO₄·7H₂O
Bod tání: 735 °C
Hustota: 2,03
Storage Temperature: Ambient
MDL Number: MFCD00149658
CAS číslo: 10026-24-1
EINECS: 233-334-2
UN: 3077
ADR: 9,III
Merck Index: 14,2448

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR