Tisk…

Oxid kobaltnato-kobaltitý ≥99.9985% (metals basis), Puratronic®

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Tricobalt tetraoxide, Cobaltic tetraoxide

10695.09 10695.18
10695.09EA 4470 CZK
10695.09 10695.18
Oxid kobaltnato-kobaltitý ≥99.9985% (metals basis), Puratronic®
Oxid kobaltnato-kobaltitý

Purity: 99.9985% CAS No.: 1308-06-1 Molecular Formula: Co3O4 Formula Weight: 240.80 Physical Form: Powder Density: 6.11 Appearance: Black Melting Point: 900° dec. Merck Reference: 14,2429 UN No.: 3077 MDL No.: MFCD00010939

Vzorec: Co₃O₄
Hustota: 6,11
Storage Temperature: Ambient
MDL Number: MFCD00010939
CAS číslo: 1308-06-1
EINECS: 215-157-2
UN: 3077
ADR: 9,III
Merck Index: 14,2429

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR