Tisk…

Fosforyltrichlorid ≥99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Phosphorus (V) oxide chloride, Phosphorus oxychloride, Phosphoryl chloride, Phosphorus(V) oxychloride

10525.30
10525.30EA 1990 CZK
10525.30
Fosforyltrichlorid ≥99%
Fosforyltrichlorid

Purity: 99% CAS No.: 10025-87-3 Molecular Formula: Cl3OP Formula Weight: 153.33 Physical Form: Liquid Density: 1.645 Refractive Index: 1.460 Sensitivity: Moisture Sensitive Appearance: Colourless Odour: Pungent Melting Point: 1.25° Boiling Point: 105.8° Merck Reference: 14,7349 UN No.: 1810 MDL No.: MFCD00011443

Vzorec: POCl₃
MW: 153.33
Bod varu: 105.8 °C
Bod tání: 1.25 °C
Hustota: 1.645
Storage Temperature: Ambient
MDL Number: MFCD00011443
CAS číslo: 10025-87-3
EINECS: 233-046-7
UN: 1810
ADR: 6.1 I
Merck Index: 12,07506

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR