Tisk…

3-Aminophenylboronic acid hemisulphate 98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: (3-Aminophenyl)boronic acid sulphate (2:1), 3-Aminobenzeneboronic acid hemisulphate

104300050 104300250
104300050.EA 4010 CZK
104300050. 104300250.
3-Aminophenylboronic acid hemisulphate 98%
3-Aminophenylboronic acid hemisulphate
Vzorec: C₁₂H₁₈B₂N₂O₈S
Storage Temperature: Refrigerator
MDL Number: MFCD00013111
CAS číslo: 66472-86-4

Objednat

Výsledky specifikačního testu

Appearance White to beige to grey to light brown Powder or crystals
Infrared spectrum Conforms
Titration with NaOH 97.5 to 102.5 %
Fluoride (F) ≤50 ppm
Arsenic (As) ≤3 ppm
Barium (Ba) Passes test
Iron (Fe) ≤0.1 %
Lead (Pb) ≤3 ppm
Mercury (Hg) ≤0.5 ppm
Acid insoluble substances ≤0.2 %
Magnesium & Alkali Salts ≤1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR