Tisk…

Tantalum ≥99.95% (metals basis), foil, Thickness: 2.0 mm (0.08 in)

Dodavatel: Thermo Fisher Scientific

10357.FF 10357.FI 10357.GH
10357.FFEA 6630 CZK
10357.FF 10357.FI 10357.GH
Tantalum ≥99.95% (metals basis), foil, Thickness: 2.0 mm (0.08 in)
Tantalum

Purity: 99.95% CAS No.: 7440-25-7 Physical Form: ≈20.8g/25x25mm Odour: Odourless MDL No.: MFCD00011252

Vzorec: Ta
MW: 180.95
Bod varu: 5427
Bod tání: 2996
Hustota: 16.69 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011252
CAS číslo: 7440-25-7
EINECS: 231-135-5
Merck Index: 12,09223

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR