Tisk…

Tris-(hydroxymethyl)aminomethan 99,8-100,5, AnalaR NORMAPUR® zA

Dodavatel: VWR Chemicals
Varování

Synonyms: 2-Amino-2-(hydroxymethyl)propane-1,3-diol(tris), Tromethamine, Tris(hydroxymethyl)methylamine, 2-Amino-2-(hydroxymethyl)propane-1,3-diol, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, Trizma(R) base, TRIS base, THAM

103154MEA 2890 CZK
103154M 103156X 103157P 10315AC
Tris-(hydroxymethyl)aminomethan 99,8-100,5, AnalaR NORMAPUR® zA
Tris-(hydroxymethyl)aminomethan
Vzorec: NH₂C(CH₂OH)₃
MW: 121.14
Bod varu: 357
Bod tání: 169…172 °C
Hustota: 1.328 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004679
CAS číslo: 77-86-1
EINECS: 201-064-4
Merck Index: 13,09842
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11a03d1ab6b2c11b9a2c83f0a73fc4264cec0110&vid=5ce890cdf298a59a7900a07fee1c857a5e5e61e0&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Assay (calculated on anhydrous) 99.8 - 100.5 %
Appearance White crystalline powder
Identification Passes test
Heavy metals (as Pb) Max. 5 ppm
Ignition residue (SO4) Max. 100 ppm
Insolubility in water Max. 30 ppm
Water Max. 0.1 %
Cl (Chloride) Max. 5 ppm
SO4 (Sulphate) Max. 5 ppm
As (Arsenic) Max. 0.2 ppm
Ca (Calcium) Max. 10 ppm
Cd (Cadmium) Max. 0.1 ppm
Cu (Copper) Max. 0.1 ppm
Fe (Iron) Max.1 ppm
K (Potassium) Max. 1 ppm
Mg (Magnesium) Max. 0.3 ppm
Na (Sodium) Max. 5 ppm
Pb (Lead) Max. 0.1 ppm
Absorbance (280 nm) (0.5 mol/l) Max. 0.05


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR