Tisk…

Rhenium ≥99.97% (metals basis), foil, Thickness: 0.025 mm (0.001 in)

Dodavatel: Thermo Fisher Scientific

10307.FF 10307.FI
10307.FFEA 8450 CZK
10307.FF 10307.FI
Rhenium ≥99.97% (metals basis), foil, Thickness: 0.025 mm (0.001 in)
Rhenium

Purity: 99.97% CAS No.: 7440-15-5 Physical Form: ≈0.33g/25x25mm Odour: Odourless MDL No.: MFCD00011195

Vzorec: Re
Bod varu: 5630 °C (1013 hPa)
Bod tání: 3180 °C
Hustota: 21,02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011195
CAS číslo: 7440-15-5
EINECS: 231-124-5
Merck Index: 12,08343

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR