Tisk…

Platina ≥99.9% (metals basis), gauze woven from wire 52 mesh, Ø 0.1 mm (0.004 in)

Dodavatel: Thermo Fisher Scientific

Synonyms: Platinium

10283.FF 10283.FI 10283.GH 10283.FL
10283.FFEA 9200 CZK
10283.FF 10283.FI 10283.GH 10283.FL
Platina ≥99.9% (metals basis), gauze woven from wire 52 mesh, Ø 0.1 mm (0.004 in)
Platina

Purity: 99.9% CAS No.: 7440-06-4 Physical Form: ≈0.47g/25x25mm, Wire Cloth Odour: Odourless MDL No.: MFCD00011179

Vzorec: Pt
Bod varu: 3827 °C (1013 hPa)
Bod tání: 1769 °C
Hustota: 21,447 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS číslo: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR