Tisk…

hořčík ≥99.8% (metals basis), turnings

Dodavatel: Thermo Fisher Scientific
Nebezpečí

10235.22 10235.36
10235.22EA 1960 CZK
10235.22 10235.36
hořčík ≥99.8% (metals basis), turnings
hořčík

Purity: 99.8% Physical Form: 2.3mm (0.09in) wide Appearance: Silver-grey Odour: Odourless UN No.: 1869 MDL No.: MFCD00085308

Vzorec: Mg
Storage Temperature: Ambient
MDL Number: MFCD00085308
CAS číslo: 7439-95-4
EINECS: 231-104-6
UN: 1869
ADR: 4.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR