Tisk…

Petrolether, 100…120 °C, AnalaR NORMAPUR® ACS zA

Dodavatel: VWR Chemicals
Nebezpečí

Synonyms: Gasoline, Petroleum ether, Petrol, Benzine

101814REA 3660 CZK
101814R
Petrolether, 100…120 °C, AnalaR NORMAPUR® ACS zA
Ropný líh
Bod varu: 100…120 °C (1013 hPa)
Hustota: 0.7 g/cm³ (20 °C)
Bod vzplanutí: –10 °C
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS číslo: 64742-49-0
EINECS: 265-151-9
UN: 1268
ADR: 3 II

Objednat

Výsledky specifikačního testu

Acidity Max. 0.00005 meq/g
Aromatics (as C₆H₆) Max. 20 ppm
Bromine value Max. 3.0
Colouration Max. 10 APHA
Distillation (min. 90 %) 100 -- 120 °C
Evaporation residue Max. 10 ppm
Substances coloured by H₂SO₄ Max. 10 APHA
Sulphur compounds (as S) Max. 50 ppm
Water Max. 100 ppm
Al (Aluminium) Max. 0.1 ppm
Ba (Barium) Max. 0.05 ppm
Ca (Calcium) Max. 0.5 ppm
Cd (Cadmium) Max. 0.05 ppm
Co (Cobalt) Max. 0.02 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.02 ppm
Fe (Iron) Max. 0.1 ppm
K (Potassium) Max. 0.2 ppm
Mg (Magnesium) Max. 0.05 ppm
Mn (Manganese) Max. 0.02 ppm
Na (Sodium) Max. 0.5 ppm
Ni (Nickel) Max. 0.02 ppm
Pb (Lead) Max. 0.05 ppm
Sn (Tin) Max. 0.1 ppm
Sr (Strontium) Max. 0.02 ppm
Zn (Zinc) Max. 0.1 ppm
Conforms to ACS Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR