Tisk…

Kadmium ≥99.99% (metals basis), rods, Ø 12.7 mm (0.5 in)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

10119.UI 10119.UT
10119.UIEA 9440 CZK
10119.UI 10119.UT
Kadmium ≥99.99% (metals basis), rods, Ø 12.7 mm (0.5 in)
Kadmium

Purity: 99.99+% CAS No.: 7440-43-9 Physical Form: ≈11.0g/cm MDL No.: MFCD00010914

Vzorec: Cd
Bod varu: 765 °C (1013 hPa)
Bod tání: 321 °C
Hustota: 8,642 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010914
CAS číslo: 7440-43-9
EINECS: 231-152-8
UN: 2570
ADR: 6.1,II
Merck Index: 12,01649

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR