Tisk…

D(-)-Luciferin potassium salt

Dodavatel: Biotium

Synonyms: (S)-2-(6-Hydroxy-2-benzothiazolyl)-2-thiazoline-4-carboxylic acid potassium salt, Beetle Luciferin potassium salt, D-Luciferin potassium, Luciferin potassium salt, Firefly luciferin potassium salt, Potassium (S)-2-(6-hydroxybenzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylate

10101-2 10101 10101-1
10101-2EA 33090 CZK
10101-2 10101. 10101-1
D(-)-Luciferin potassium salt
D-(-)-Luciferin potassium salt
Vzorec: C₁₁H₇KN₂O₃S₂
MW: 318.41 g/mol
MDL Number: MFCD00044928
CAS číslo: 115144-35-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR