Tisk…

Ethanolamin, EMPROVE® EXPERT BP, NF BiopharmaceuticalProduction, SAFC®

Dodavatel: MERCK PRODUCTION CHEMICALS
Nebezpečí

Synonyms: 2-Aminoethanol, Monoethanolamine

1.37044.9025
1.37044.9025EA 0 CZK
1.37044.9025
Ethanolamin, EMPROVE® EXPERT BP, NF BiopharmaceuticalProduction, SAFC®
Ethanolamin
Vzorec: NH₂CH₂CH₂OH
MW: 61.08
Bod varu: 171
Bod tání: 10.5
Hustota: 1.0164 g/cm³ (20 °C)
Bod vzplanutí: 93
Storage Temperature: Ambient
MDL Number: MFCD00008183
CAS číslo: 141-43-5
EINECS: 205-483-3
UN: 2491
ADR: 8
Merck Index: 13,03762
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82bfb6ea7cf5dde341c60ff9cf752bc5c422eb28&vid=b387e8c39029da4ea050ba6dcfb6123d81ab03c8&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR