Tisk…
Test kits, cyanuric acid, Spectroquant®, Supelco®
Dodavatel: Merck
1.19253.0001
1.19253.0001EA 2660 CZK
1.19253.0001
Test kits, cyanuric acid, Spectroquant®, Supelco®
Kity pro environmentální testy
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
  • Highly stable, ready to use reagent mixtures in a user friendly design
  • Free selection of appropriate cell format according to the measuring range
  • Long shelf life up to 3 years - all at room temperature
  • Automatic selection of the correct method with help of AutoSelector barcode identification
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.