Tisk…

Voda, EMSURE® analysis_for, Supelco®

Dodavatel: Merck

Synonyms: Hydrogen oxide

MQ300

1.16754.5000 1.16754.9010 1.16754.4000
1.16754.5000EA 1530 CZK
1.16754.5000 1.16754.9010 1.16754.4000
Voda, EMSURE® analysis_for, Supelco®
Voda

EMSURE® grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE® provides worldwide best and most extensive product specifications. Merck declare the EMSURE® range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.

Vzorec: H₂O
MW: 18.02
Bod varu: 100
Bod tání: 0
Hustota: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS číslo: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Objednat

Výsledky specifikačního testu

Molar Mass 18.02 g/mol
Boiling Point 100 °C (1013 hPa)
Vapor Pressure 23 hPa (20 °C)
Density 1.00 g/cm3 (20 °C)
pH (H2O, 20 °C) neutral


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR